Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lztr1
Entrez Gene ID
66863
Gene Name
leucine-zipper-like transcriptional regulator, 1
Gene Aliases
1200003E21Rik, AI591627, AW550890
Chromosome Location
Chr.16: 17508541 - 17526333 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025808.3 NP_080084.2 19 - 20 2370 82
GenBank mRNA AK004561.2 - 19 - 20 2370 82
AK005037.1 - 19 - 20 2369 82
AK083411.1 - 19 - 20 2374 82
AK088938.1 - 10 - 11 1302 82
BC034400.1 - 19 - 20 2364 82

View Full Product Details