The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017382.5 | NP_059078.2 | 1 - 2 | 178 | 68 |
GenBank mRNA | AB232605.1 | - | 1 - 2 | 45 | 68 |
AK014268.1 | - | 1 - 2 | 179 | 68 | |
AK031143.1 | - | 1 - 2 | 82 | 68 | |
AK132072.1 | - | 1 - 2 | 162 | 68 | |
AK132159.1 | - | 1 - 2 | 164 | 68 | |
AK147122.1 | - | 1 - 2 | 179 | 68 | |
BC010722.1 | - | 1 - 2 | 167 | 68 |