Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eepd1
Entrez Gene ID
67484
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
2310005P05Rik, AI481005
Chromosome Location
Chr.9: 25481526 - 25604110 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026189.3 NP_080465.3 1 - 2 300 94
GenBank mRNA AK009180.2 - 1 - 2 295 94
AK019086.1 - 1 - 2 295 94
AK168564.1 - 1 - 2 301 94
AK173239.1 - 1 - 2 350 94
BC025571.1 - 1 - 2 364 94
BY127756.1 - 1 - 2 301 94

View Full Product Details