The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013895.4 | NP_038923.1 | 1 - 2 | 348 | 81 |
GenBank mRNA | AF144702.1 | - | 1 - 2 | 350 | 81 |
AK003054.1 | - | 1 - 2 | 154 | 81 | |
AK009041.1 | - | 1 - 2 | 142 | 81 | |
AK009152.1 | - | 1 - 2 | 158 | 81 | |
AK013230.1 | - | 1 - 2 | 146 | 81 | |
AK088984.1 | - | 1 - 2 | 158 | 81 | |
BC011436.1 | - | 1 - 2 | 161 | 81 |