The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024910.3 | NP_001020081.2 | 1 - 2 | 459 | 72 |
NM_001024911.2 | NP_001020082.1 | 1 - 2 | 459 | 72 | |
GenBank mRNA | AK076569.1 | - | 1 - 2 | 460 | 72 |
BC053752.1 | - | 1 - 2 | 73 | 72 |