The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024910.3 | NP_001020081.2 | 8 - 9 | 1524 | 135 |
NM_001024911.2 | NP_001020082.1 | 8 - 9 | 1524 | 135 | |
GenBank mRNA | AK076569.1 | - | 8 - 9 | 1525 | 135 |
BC053752.1 | - | 8 - 9 | 1138 | 135 |