Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Prodh2
- Entrez Gene ID
-
56189
- Gene Name
- proline dehydrogenase (oxidase) 2
- Gene Aliases
-
2510028N04Rik, 2510038B11Rik, MmPOX, MmPOX1, POX1
- Chromosome Location
-
Chr.7: 30493586 - 30513402 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK011039.1 |
- |
7 - 8 |
1031 |
76 |
View Full Product Details