Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem26
Entrez Gene ID
327766
Gene Name
transmembrane protein 26
Gene Aliases
3230401A06, AI195372
Chromosome Location
Chr.10: 68723746 - 68782655 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177794.3 NP_808462.2 4 - 5 753 81
GenBank mRNA AK028283.1 - 4 - 5 754 81
AK084351.1 - 4 - 5 731 81
AK143866.1 - 4 - 5 731 81
BC117502.1 - 5 - 6 757 81
BC117503.1 - 4 - 5 675 81

View Full Product Details