Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Micu1
Entrez Gene ID
216001
Gene Name
mitochondrial calcium uptake 1
Gene Aliases
C730016L05Rik, Calc, Cbara1
Chromosome Location
Chr.10: 59702482 - 59864134 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291442.1 NP_001278371.1 9 - 10 1223 104
NM_001291443.1 NP_001278372.1 10 - 11 1217 104
NM_144822.3 NP_659071.1 9 - 10 1205 104
GenBank mRNA AK046049.1 - 9 - 10 1188 104
AK049521.1 - 9 - 10 1201 104
AK050113.1 - 9 - 10 1192 104
AK052485.1 - 9 - 10 1153 104
AK077605.1 - 9 - 10 1155 104
AK085553.1 - 9 - 10 1124 105
BC018320.1 - 9 - 10 1146 104
BC023022.1 - 10 - 11 1152 104
BC023042.1 - 8 - 9 1089 104
BC026566.1 - 9 - 10 1156 104

View Full Product Details