Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtf2a2
Entrez Gene ID
235459
Gene Name
general transcription factor II A, 2
Gene Aliases
TFIIA-gamma, TfIIg, Tfiia2
Chromosome Location
Chr.9: 70011851 - 70022866 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_104093.1 3 - 4 337 60
GenBank mRNA BC028551.1 - 3 - 4 319 60

View Full Product Details