Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abcf3
Entrez Gene ID
27406
Gene Name
ATP-binding cassette, sub-family F (GCN20), member 3
Gene Aliases
AI326318, AU016058, BB119416
Chromosome Location
Chr.16: 20548556 - 20561603 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013852.2 NP_038880.1 9 - 10 1051 83
GenBank mRNA AK030626.1 - 9 - 10 1016 83
AK179781.1 - 3 - 4 83
AK181390.1 - 1 - 2 47 83
AK211939.1 - 1 - 2 47 83
BC032923.1 - 9 - 10 992 83

View Full Product Details