Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd3g
Entrez Gene ID
12502
Gene Name
CD3 antigen, gamma polypeptide
Gene Aliases
Ctg-3, Ctg3, T3g
Chromosome Location
Chr.9: 44969572 - 44980431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009850.2 NP_033980.1 4 - 5 555 75
GenBank mRNA AK134049.1 - 4 - 5 488 75
AK153980.1 - 4 - 5 487 75
AK153987.1 - 4 - 5 474 75
BC027528.1 - 4 - 5 486 75
EF474453.1 - 4 - 5 444 75
GU456630.1 - 4 - 5 444 75
M58149.1 - 4 - 5 461 75
Y00635.1 - 4 - 5 583 75

View Full Product Details