Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc84
Entrez Gene ID
382073
Gene Name
coiled-coil domain containing 84
Gene Aliases
D630044F24Rik, Gm1114
Chromosome Location
Chr.9: 44410159 - 44418084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201372.3 NP_958760.2 6 - 7 600 69
GenBank mRNA AK052757.1 - 6 - 7 643 69
BC055929.1 - 6 - 7 600 69
BC096560.1 - 6 - 7 605 69
BC116877.1 - 6 - 7 642 69
BC116879.1 - 6 - 7 642 69

View Full Product Details