The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175481.5 | NP_780690.2 | 20 - 21 | 2710 | 135 |
GenBank mRNA | AK032029.1 | - | 20 - 21 | 2711 | 135 |
AK080582.1 | - | 20 - 21 | 2840 | 135 | |
BC118010.2 | - | 19 - 20 | 2625 | 135 |