Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtkn2
Entrez Gene ID
170799
Gene Name
rhotekin 2
Gene Aliases
B130039D23Rik, Mbf, Plekhk1
Chromosome Location
Chr.10: 67979566 - 68043864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK052662.1 - 3 - 4 506 106
BC116341.2 - - 470 106
BC116342.2 - - 470 106
BC158093.1 - 3 - 4 473 106

View Full Product Details