The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164147.1 | NP_001157619.1 | 16 - 17 | 1845 | 73 |
NM_001164148.1 | NP_001157620.1 | 16 - 17 | 1842 | 73 | |
NM_001164149.1 | NP_001157621.1 | 16 - 17 | 1839 | 73 | |
NM_001164150.1 | NP_001157622.1 | 16 - 17 | 1845 | 73 | |
NM_001164151.1 | NP_001157623.1 | 16 - 17 | 1842 | 73 | |
NM_001164152.1 | NP_001157624.1 | 16 - 17 | 1842 | 73 | |
NM_001164153.1 | NP_001157625.1 | 16 - 17 | 1842 | 73 | |
NM_011548.4 | NP_035678.3 | 16 - 17 | 1839 | 73 | |
GenBank mRNA | AF352579.1 | - | 16 - 17 | 1534 | 73 |
AK017617.1 | - | 16 - 17 | 1807 | 73 | |
AK034777.1 | - | 16 - 17 | 1825 | 73 | |
AK038738.1 | - | 16 - 17 | 1829 | 73 | |
AK146786.1 | - | 16 - 17 | 1837 | 73 | |
AK147735.1 | - | 15 - 16 | 1502 | 73 | |
AK156265.1 | - | 16 - 17 | 1822 | 73 | |
AK161009.1 | - | 16 - 17 | 1831 | 73 | |
AK170674.1 | - | 16 - 17 | 1837 | 73 | |
BC006860.1 | - | 16 - 17 | 1793 | 73 | |
BC018260.1 | - | 15 - 16 | 1467 | 73 | |
D16631.1 | - | 2 - 3 | 294 | 73 | |
D16632.1 | - | 2 - 3 | 294 | 73 | |
D16633.1 | - | 2 - 3 | 294 | 73 | |
D16634.1 | - | 2 - 3 | 294 | 73 | |
D16635.1 | - | 2 - 3 | 297 | 73 | |
D16636.1 | - | 2 - 3 | 294 | 73 | |
D16637.1 | - | 2 - 3 | 294 | 73 | |
D16638.1 | - | 2 - 3 | 294 | 73 | |
X17500.1 | - | 3 - 4 | 347 | 73 |