Assay Design
The assay probe spans an exon junction.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK006740.1 |
- |
|
142 |
|
Chr. - - [-] on Build |
73 |
GenBankmRNA |
BY706724.1 |
- |
2 - 3 |
142 |
|
Chr. 9 - 7965838 - 7977004 [+] on Build GRCm38 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
yes-associated protein 1 |
Yap1 |
AI325207, Yap, Yap65, Yki, Yorkie |
|
Anti-sense |
Chr. - 7931999 - 8004596 on Build GRCm38 |
Downstream |
RIKEN cDNA 9230110C19 gene |
9230110C19Rik |
|
47.6kb
|
Anti-sense |
Chr. - 8021672 - 8042894 on Build GRCm38 |
View Full Product Details