The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144923.3 | NP_659172.1 | 1 - 2 | 220 | 73 |
GenBank mRNA | AK153153.1 | - | 1 - 2 | 141 | 73 |
BC006617.1 | - | 1 - 2 | 160 | 73 | |
BC027279.1 | - | 1 - 2 | 220 | 73 |