The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001043317.2 | NP_001036782.1 | 3 - 4 | 346 | 62 |
NR_102722.1 | 3 - 4 | 346 | 62 | ||
NR_102723.1 | 3 - 4 | 360 | 62 | ||
GenBank mRNA | AK041230.1 | - | 3 - 4 | 281 | 62 |
AK143835.1 | - | 3 - 4 | 284 | 62 | |
BC046957.1 | - | 3 - 4 | 272 | 62 | |
BY231006.1 | - | 3 - 4 | 284 | 62 | |
L02844.1 | - | 2 - 3 | 492 | 62 | |
L16928.1 | - | 2 - 3 | 487 | 62 |