Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nfkbid
Entrez Gene ID
243910
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, delta
Gene Aliases
IkappaBNS
Chromosome Location
Chr.7: 30421729 - 30428746 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172142.3 NP_742154.1 1 - 2 199 152
GenBank mRNA AK155969.1 - 2 - 3 205 152
AK167356.1 - 2 - 3 193 152
AY078069.1 - 1 - 2 204 152
BC110633.1 - 1 - 2 159 152

View Full Product Details