The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199494.1 | NP_001186423.1 | 10 - 11 | 1106 | 62 |
NM_007915.5 | NP_031941.1 | 10 - 11 | 990 | 62 | |
GenBank mRNA | AK145018.1 | - | 10 - 11 | 1068 | 62 |
AK147776.1 | - | 10 - 11 | 1019 | 62 | |
AK159927.1 | - | 10 - 11 | 1048 | 62 | |
AK172233.1 | - | 10 - 11 | 1070 | 62 | |
U41751.2 | - | 10 - 11 | 994 | 62 |