Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cplx3
Entrez Gene ID
235415
Gene Name
complexin 3
Gene Aliases
BC016632, CpxIII, ERGIC-53L, ERGL, Lamn1l, Lman1l
Chromosome Location
Chr.9: 57599992 - 57606281 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146223.3 NP_666335.1 1 - 2 302 69
GenBank mRNA AK044211.1 - 1 - 2 267 69
AY264290.1 - 1 - 2 244 69
BC016632.1 - 1 - 2 251 69
BC024854.1 - 1 - 2 252 69
BC033343.1 - 1 - 2 253 69

View Full Product Details