Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zc3h12c
- Entrez Gene ID
-
244871
- Gene Name
- zinc finger CCCH type containing 12C
- Gene Aliases
-
A230108E06, C230027N18Rik, mKIAA1726
- Chromosome Location
-
Chr.9: 52109583 - 52169154 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC042784.1 |
- |
3 - 4 |
|
76 |
View Full Product Details