Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h12c
Entrez Gene ID
244871
Gene Name
zinc finger CCCH type containing 12C
Gene Aliases
A230108E06, C230027N18Rik, mKIAA1726
Chromosome Location
Chr.9: 52109583 - 52169154 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162921.1 NP_001156393.1 4 - 5 1209 103
GenBank mRNA AK220416.1 - 2 - 3 339 103

View Full Product Details