The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281954.1 | NP_001268883.1 | 6 - 7 | 782 | 112 |
NM_019465.4 | NP_062338.3 | 6 - 7 | 782 | 112 | |
GenBank mRNA | AF001104.1 | - | 6 - 7 | 728 | 112 |
AK005719.1 | - | 4 - 5 | 505 | 112 | |
AK163744.1 | - | 1 - 2 | 180 | 112 | |
BC117715.1 | - | 6 - 7 | 727 | 112 | |
BC117716.1 | - | 6 - 7 | 728 | 112 |