Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmprss9
Entrez Gene ID
432478
Gene Name
transmembrane protease, serine 9
Gene Aliases
Serase-1B
Chromosome Location
Chr.10: 80871117 - 80899495 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081688.2 NP_001075157.2 15 - 16 2735 68
GenBank mRNA BC158102.1 - 15 - 16 2831 68
BC172169.1 - 15 - 16 2831 68

View Full Product Details