The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081688.2 | NP_001075157.2 | 7 - 8 | 846 | 112 |
GenBank mRNA | AB109391.1 | - | 7 - 8 | 957 | 112 |
BC158102.1 | - | 7 - 8 | 942 | 112 | |
BC172169.1 | - | 7 - 8 | 942 | 112 |