Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prtg
Entrez Gene ID
235472
Gene Name
protogenin homolog (Gallus gallus)
Gene Aliases
A230098A12Rik, Igdcc5
Chromosome Location
Chr.9: 72807274 - 72917307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175485.4 NP_780694.3 6 - 7 1142 87
GenBank mRNA AK083540.1 - 4 - 5 578 87
AY630257.1 - 6 - 7 1212 87
BC116336.1 - 4 - 5 575 87
BC116337.1 - 4 - 5 575 87
DQ360114.1 - 6 - 7 1142 87

View Full Product Details