The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045518.1 | NP_001038983.1 | 3 - 4 | 751 | 78 |
GenBank mRNA | AK049628.1 | - | 4 - 5 | 843 | 78 |
AK145108.1 | - | 4 - 5 | 835 | 78 | |
BC120577.1 | - | 3 - 4 | 751 | 78 | |
BC120579.1 | - | 3 - 4 | 751 | 78 |