Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elmod1
Entrez Gene ID
270162
Gene Name
ELMO/CED-12 domain containing 1
Gene Aliases
4831417L10, AI428506, AI834991
Chromosome Location
Chr.9: 53911457 - 53975310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177769.4 NP_808437.2 9 - 10 1034 182
GenBank mRNA AK029207.1 - 9 - 10 915 182
AK132239.1 - 9 - 10 1035 182
AK164094.1 - - 186 182
BC120566.1 - 8 - 9 750 182
BC144961.1 - 8 - 9 750 182
BC144963.1 - 8 - 9 712 182

View Full Product Details