Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc36a4
Entrez Gene ID
234967
Gene Name
solute carrier family 36 (proton/amino acid symporter), member 4
Gene Aliases
6330573I15Rik, PAT4
Chromosome Location
Chr.9: 15709769 - 15742388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172289.4 NP_758493.2 5 - 6 474 62
GenBank mRNA AK032074.1 - 5 - 6 475 62
AK052277.1 - 5 - 6 475 62
AK150167.1 - 5 - 6 506 62
BC115964.1 - 5 - 6 475 62

View Full Product Details