Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc36a4
Entrez Gene ID
234967
Gene Name
solute carrier family 36 (proton/amino acid symporter), member 4
Gene Aliases
6330573I15Rik, PAT4
Chromosome Location
Chr.9: 15709769 - 15742388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172289.4 NP_758493.2 6 - 7 559 91
GenBank mRNA AF453746.1 - 7 - 8 558 91
AK032074.1 - 6 - 7 560 91
AK052277.1 - 6 - 7 560 91
AK150167.1 - 6 - 7 591 91
BC115964.1 - 6 - 7 560 91

View Full Product Details