Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aagab
Entrez Gene ID
66939
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
2310007F21Rik, p34
Chromosome Location
Chr.9: 63602629 - 63641897 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025857.2 NP_080133.1 6 - 7 660 73
GenBank mRNA AK159897.1 - 6 - 7 661 73
AK209565.1 - 1 - 2 34 73
BC027311.1 - 6 - 7 656 73

View Full Product Details