Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid3b
Entrez Gene ID
56380
Gene Name
AT rich interactive domain 3B (BRIGHT-like)
Gene Aliases
Bdp, Dri2
Chromosome Location
Chr.9: 57790353 - 57836847 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019689.2 NP_062663.1 6 - 7 1517 81
GenBank mRNA AF116847.1 - 6 - 7 1517 81
AK084952.1 - 4 - 5 838 81
AK148073.1 - 7 - 8 1615 81
BC145330.1 - 6 - 7 1581 81
BC150755.1 - 6 - 7 1511 81

View Full Product Details