Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d21
Entrez Gene ID
74286
Gene Name
TBC1 domain family, member 21
Gene Aliases
1700095K08Rik, MgcRabGAP
Chromosome Location
Chr.9: 58359714 - 58370479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028854.3 NP_083130.1 3 - 4 319 80
GenBank mRNA AK007086.1 - 3 - 4 407 80
BC100468.1 - 3 - 4 385 80
BC132329.1 - 3 - 4 319 80
BC138332.1 - 3 - 4 319 80
BC145239.1 - 3 - 4 319 80
BC145240.1 - 3 - 4 319 80

View Full Product Details