Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stra6
Entrez Gene ID
20897
Gene Name
stimulated by retinoic acid gene 6
Gene Aliases
AI891933
Chromosome Location
Chr.9: 58063793 - 58154187 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162475.1 NP_001155947.1 9 - 10 864 92
NM_001162476.1 NP_001155948.1 9 - 10 1175 92
NM_001162479.1 NP_001155951.1 9 - 10 969 92
NM_009291.2 NP_033317.2 9 - 10 1134 92
GenBank mRNA AF062476.1 - 9 - 10 1134 92
BC075657.1 - 9 - 10 864 92
CX237908.1 - 7 - 8 661 92

View Full Product Details