The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025375.2 | NP_001020546.1 | 6 - 7 | 690 | 138 |
NM_001025376.2 | NP_001020547.1 | 6 - 7 | 579 | 138 | |
NM_023191.3 | NP_075680.1 | 6 - 7 | 584 | 138 | |
GenBank mRNA | AF309554.1 | - | 6 - 7 | 542 | 138 |
AK010283.1 | - | 6 - 7 | 530 | 138 | |
AK012342.1 | - | 6 - 7 | 596 | 138 | |
AK013114.1 | - | 6 - 7 | 548 | 138 | |
AK077037.1 | - | - | 624 | 138 | |
AK142498.1 | - | 6 - 7 | 547 | 138 | |
AK153862.1 | - | 6 - 7 | 617 | 138 | |
AK167706.1 | - | 6 - 7 | 617 | 138 | |
BC023026.1 | - | 6 - 7 | 529 | 138 | |
EF011623.1 | - | 6 - 7 | 527 | 138 |