Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oscp1
Entrez Gene ID
230751
Gene Name
organic solute carrier partner 1
Gene Aliases
1810007P19Rik, 5730415O04, 6030436A01Rik
Chromosome Location
Chr.4: 126057287 - 126089334 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172701.2 NP_766289.2 2 - 3 386 66
GenBank mRNA AB154826.1 - 2 - 3 339 66
AB286960.1 - 3 - 4 387 66
AK077467.1 - 2 - 3 352 66
AK077909.1 - 2 - 3 307 66
BC045150.1 - 2 - 3 332 66

View Full Product Details