Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmym6
Entrez Gene ID
100177
Gene Name
zinc finger, MYM-type 6
Gene Aliases
9330177P20Rik, AI593204, AI661340, D4Wsu24e, Zfp258
Chromosome Location
Chr.4: 127077383 - 127124374 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC140958.1 - 9 - 10 1383 61
BC144807.1 - 9 - 10 1390 61

View Full Product Details