Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sptbn1
Entrez Gene ID
20742
Gene Name
spectrin beta, non-erythrocytic 1
Gene Aliases
9930031C03Rik, AL033301, SPTB2, Spnb-2, Spnb2, elf1, elf3, mKIAA4049
Chromosome Location
Chr.11: 30099395 - 30268204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009260.2 NP_033286.2 11 - 12 2030 113
NM_175836.2 NP_787030.2 11 - 12 1690 113
GenBank mRNA AF017112.1 - 11 - 12 1937 113
AK157727.1 - 11 - 12 2020 113
AK220456.1 - 12 - 13 1892 113
M74773.1 - 11 - 12 2010 113

View Full Product Details