Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sptbn1
Entrez Gene ID
20742
Gene Name
spectrin beta, non-erythrocytic 1
Gene Aliases
9930031C03Rik, AL033301, SPTB2, Spnb-2, Spnb2, elf1, elf3, mKIAA4049
Chromosome Location
Chr.11: 30099395 - 30268204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009260.2 NP_033286.2 8 - 9 1452 81
NM_175836.2 NP_787030.2 8 - 9 1112 81
GenBank mRNA AK157727.1 - 8 - 9 1442 81
AK169544.1 - 8 - 9 1452 81
AK220456.1 - 9 - 10 1314 81
M74773.1 - 8 - 9 1432 81

View Full Product Details