Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm4a
Entrez Gene ID
230674
Gene Name
lysine (K)-specific demethylase 4A
Gene Aliases
D4Ertd222e, JHDM3A, Jmjd2, Jmjd2a, mKIAA0677
Chromosome Location
Chr.4: 118136957 - 118180043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161823.1 NP_001155295.1 10 - 11 1554 108
NM_172382.2 NP_759014.2 10 - 11 1554 108
GenBank mRNA AK054095.1 - 10 - 11 1518 108
AK129187.1 - 10 - 11 1409 108
AK136085.1 - 1 - 2 211 108
AK145066.1 - 10 - 11 1540 108
AK145947.1 - 10 - 11 1476 108
BC028866.1 - 10 - 11 1457 108

View Full Product Details