The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133872.2 | NP_598633.2 | 14 - 15 | 1934 | 60 |
GenBank mRNA | AK007763.1 | - | 1 - 2 | 142 | 60 |
AK129170.1 | - | 14 - 15 | 1874 | 60 | |
BC059885.1 | - | 14 - 15 | 1921 | 60 |