The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146153.3 | NP_666265.3 | 9 - 10 | 2644 | 74 |
GenBank mRNA | AK046010.1 | - | 1 - 2 | 977 | 74 |
AK153954.1 | - | 3 - 4 | 443 | 74 | |
BC033472.1 | - | 1 - 2 | 241 | 74 | |
BC040346.1 | - | 9 - 10 | 2575 | 74 | |
BC092239.1 | - | 9 - 10 | 2560 | 74 |