Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hormad2
Entrez Gene ID
75828
Gene Name
HORMA domain containing 2
Gene Aliases
4930529M09Rik
Chromosome Location
Chr.11: 4345801 - 4442459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK015939.1 - 5 - 6 503 125

View Full Product Details