The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029458.1 | NP_083734.1 | 6 - 7 | 561 | 73 |
GenBank mRNA | AK015939.1 | - | 7 - 8 | 563 | 73 |
BC120781.1 | - | 6 - 7 | 561 | 73 | |
BC120783.1 | - | 6 - 7 | 561 | 73 | |
BC144842.1 | - | 6 - 7 | 561 | 73 |