Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hormad2
Entrez Gene ID
75828
Gene Name
HORMA domain containing 2
Gene Aliases
4930529M09Rik
Chromosome Location
Chr.11: 4345801 - 4442459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029458.1 NP_083734.1 8 - 9 660 115
GenBank mRNA AK015939.1 - 9 - 10 662 115
BC120781.1 - 8 - 9 660 115
BC120783.1 - 8 - 9 660 115
BC144842.1 - 8 - 9 660 115

View Full Product Details