Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcmf1
Entrez Gene ID
74287
Gene Name
potassium channel modulatory factor 1
Gene Aliases
1700094M07Rik, Debt91, Pmcf
Chromosome Location
Chr.6: 72841114 - 72899979 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019715.2 NP_062689.2 3 - 4 676 93
GenBank mRNA AK007070.1 - 3 - 4 623 93
AK142436.1 - 3 - 4 433 93
BC029325.1 - 3 - 4 498 93
BC043330.1 - 3 - 4 457 93

View Full Product Details