The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177412.1 | NP_001170883.1 | 4 - 5 | 1037 | 59 |
GenBank mRNA | AK220266.1 | - | 4 - 5 | 611 | 59 |