The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146014.3 | NP_666126.1 | 2 - 3 | 309 | 70 |
GenBank mRNA | AK155145.1 | - | 2 - 3 | 283 | 70 |
AY442689.1 | - | 2 - 3 | 196 | 70 | |
BC029157.1 | - | 2 - 3 | 240 | 70 |